(Acetylmethylen)triphenylphosphoran

Lieferant: SIA ENAMINE

Synonyme: 1-(Triphenylphosphoranylidene)propan-2-one, 1-Triphenylphosphoranylidene-2-propanone

EN300102571.1G
ENAMEN300102571.1GEA 0 EUR
ENAMEN300102571.1G
(Acetylmethylen)triphenylphosphoran
(Acetylmethylen)triphenylphosphoran
Formel: C₂₁H₁₉OP
Molecular Weight: 318,36 g/mol
MDL: MFCD00008774
CAS-Nummer: 1439-36-7
EINECS: 215-878-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR