1,2,4,5-Tetramethyl-3,6-dinitrobenzol

Lieferant: SIA ENAMINE

Synonyme: 1,4-Dinitrodurene, Dinitrodurene

EN30010259.1G
ENAMEN30010259.1GEA 0 EUR
ENAMEN30010259.1G
1,2,4,5-Tetramethyl-3,6-dinitrobenzol
1,2,4,5-Tetramethyl-3,6-dinitrobenzol
Formel: C₁₀H₁₂N₂O₄
Molecular Weight: 224,22 g/mol
MDL: MFCD00007164
CAS-Nummer: 5465-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR