2,3-Dichlormaleinsäureanhydrid

Lieferant: SIA ENAMINE

EN300102814.1G
ENAMEN300102814.1GEA 0 EUR
ENAMEN300102814.1G
2,3-Dichlormaleinsäureanhydrid
2,3-Dichlormaleinsäureanhydrid
Formel: C₄Cl₂O₃
MDL: MFCD00005520
CAS-Nummer: 1122-17-4
EINECS: 214-343-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR