3-Chlor-5-fluorbenzoesäure

Lieferant: SIA ENAMINE
Achtung

EN300102825.1G
ENAMEN300102825.1GEA 0 EUR
ENAMEN300102825.1G
3-Chlor-5-fluorbenzoesäure
3-Chloro-5-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174.56 g/mol
MDL: MFCD00973904
CAS-Nummer: 25026-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR