2,4-Difluor-6-nitroanilin

Lieferant: SIA ENAMINE

EN300102827.1G
ENAMEN300102827.1GEA 0 EUR
ENAMEN300102827.1G
2,4-Difluor-6-nitroanilin
2,4-Difluor-6-nitroanilin
Formel: C₆H₄F₂N₂O₂
Molecular Weight: 174,11 g/mol
MDL: MFCD00034058
CAS-Nummer: 364-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR