Ethyl-5-amino-1-(3-chlorphenyl)-1H-pyrazol-4-carboxylat

Lieferant: SIA ENAMINE

Synonyme: Ethyl 5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate

EN30010285.1G
ENAMEN30010285.1GEA 0 EUR
ENAMEN30010285.1G
Ethyl-5-amino-1-(3-chlorphenyl)-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-(3-chlorphenyl)-1H-pyrazol-4-carboxylat
Formel: C₁₂H₁₂ClN₃O₂
Molecular Weight: 265,7000 g/mol
MDL: MFCD00973940
CAS-Nummer: 15001-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR