6-Methoxypicolinsäure

Lieferant: SIA ENAMINE

EN300102850.1G
ENAMEN300102850.1GEA 0 EUR
ENAMEN300102850.1G
6-Methoxypicolinsäure
6-Methoxypicolinsäure
Formel: C₇H₇NO₃
MDL: MFCD06800962
CAS-Nummer: 26893-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR