3-Brom-N,N-dimethylbenzamid

Lieferant: SIA ENAMINE

EN300103033.1G
ENAMEN300103033.1GEA 0 EUR
ENAMEN300103033.1G
3-Brom-N,N-dimethylbenzamid
3-Brom-N,N-dimethylbenzamid
Formel: C₉H₁₀BrNO
Molecular Weight: 228,0858 g/mol
MDL: MFCD01215179
CAS-Nummer: 24167-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR