(3-Iodphenyl)essigsäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 2-(3-Iodophenyl)acetic acid

EN300103230.1G
ENAMEN300103230.1GEA 0 EUR
ENAMEN300103230.1G
(3-Iodphenyl)essigsäure
(3-Iodphenyl)essigsäure
Formel: (C₆H₄I)CH₂COOH
Molecular Weight: 262,05 g/mol
MDL: MFCD00046548
CAS-Nummer: 1878-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR