Methyl-2-oxocyclopentancarboxylat

Lieferant: SIA ENAMINE

Synonyme: Methyl 2-cyclopentanonecarboxylate

EN300103267.1G
ENAMEN300103267.1GEA 0 EUR
ENAMEN300103267.1G
Methyl-2-oxocyclopentancarboxylat
Methyl-2-oxocyclopentancarboxylat
Formel: C₇H₁₀O₃
Molecular Weight: 142,16 g/mol
MDL: MFCD00001411
CAS-Nummer: 10472-24-9
EINECS: 233-962-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR