2-Brom-5-hydroxypyridin

Lieferant: SIA ENAMINE
Achtung

Synonyme: 2-Brom-5-pyridinol

EN300103371.1G
ENAMEN300103371.1GEA 0 EUR
ENAMEN300103371.1G
2-Brom-5-hydroxypyridin
2-Brom-5-hydroxypyridin
Formel: C₅H₄BrNO
MDL: MFCD04114184
CAS-Nummer: 55717-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR