4-Methoxycarbonyl-2-nitrophenylboronsäure

Lieferant: SIA ENAMINE

EN300103373.1G
ENAMEN300103373.1GEA 0 EUR
ENAMEN300103373.1G
4-Methoxycarbonyl-2-nitrophenylboronsäure
4-Methoxycarbonyl-2-nitrophenylboronsäure
Formel: C₈H₈BNO₆
MDL: MFCD01632202
CAS-Nummer: 85107-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR