4-Acetylphenylboronsäure

Lieferant: SIA ENAMINE

Synonyme: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

EN300103412.1G
ENAMEN300103412.1GEA 0 EUR
ENAMEN300103412.1G
4-Acetylphenylboronsäure
4-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163,97 g/mol
MDL: MFCD01074667
CAS-Nummer: 149104-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR