4-Brom-2-chlorpyridin

Lieferant: SIA ENAMINE

EN300103561.1G
ENAMEN300103561.1GEA 0 EUR
ENAMEN300103561.1G
4-Brom-2-chlorpyridin
4-Bromo-2-chloropyridine
Formel: C₅H₃BrClN
MDL: MFCD03840756
CAS-Nummer: 73583-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR