1-Fluor-4-ethinylbenzol

Lieferant: SIA ENAMINE

Synonyme: (4-Fluorphenyl)acetylen, 1-Ethinyl-4-fluorbenzol, (4-Fluorphenyl)ethin

EN300103797.1G
ENAMEN300103797.1GEA 0 EUR
ENAMEN300103797.1G
1-Fluor-4-ethinylbenzol
1-Fluor-4-ethinylbenzol
Formel: C₈H₅F
Molecular Weight: 120.13 g/mol
Flammpunkt: <61 °C
MDL: MFCD00168823
CAS-Nummer: 766-98-3
UN: 1325
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR