3-Chlor-5-fluoranilin

Lieferant: SIA ENAMINE

EN300103833.1G
ENAMEN300103833.1GEA 0 EUR
ENAMEN300103833.1G
3-Chlor-5-fluoranilin
3-Chlor-5-fluoranilin
Formel: C₆H₅ClFN
Molecular Weight: 145,56 g/mol
MDL: MFCD03407962
CAS-Nummer: 4863-91-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR