2-Brom-3-nitropyridin

Lieferant: SIA ENAMINE
Achtung

EN300103885.1G
ENAMEN300103885.1GEA 0 EUR
ENAMEN300103885.1G
2-Brom-3-nitropyridin
2-Bromo-3-nitropyridine
Formel: C₅H₃BrN₂O₂
Molecular Weight: 203 g/mol
MDL: MFCD00955613
CAS-Nummer: 19755-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR