2'-Brom-4'-fluoracetophenon

Lieferant: SIA ENAMINE

Synonyme: 1-(2-Bromo-4-fluorophenyl)ethanone

EN300103964.1G
ENAMEN300103964.1GEA 0 EUR
ENAMEN300103964.1G
2'-Brom-4'-fluoracetophenon
2'-Brom-4'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
MDL: MFCD00077464
CAS-Nummer: 1006-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR