2-Fluor-5-(trifluormethyl)benzonitril

Lieferant: SIA ENAMINE

Synonyme: 2-Fluor-5-(trifluormethyl)benzoesäurenitril, 3-Cyan-4-fluorbenzotrifluorid

EN300104085.1G
ENAMEN300104085.1GEA 0 EUR
ENAMEN300104085.1G
2-Fluor-5-(trifluormethyl)benzonitril
2-Fluor-5-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Molecular Weight: 189.11 g/mol
Siedepunkt: 99
MDL: MFCD00061280
CAS-Nummer: 4088-84-0
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR