5-Amino-2-fluorpyridin

Lieferant: SIA ENAMINE

Synonyme: 2-Fluorpyridin-5-amin, 2-Fluor-5-pyridylamin

EN300104103.1G
ENAMEN300104103.1GEA 0 EUR
ENAMEN300104103.1G
5-Amino-2-fluorpyridin
5-Amino-2-fluorpyridin
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
MDL: MFCD01632180
CAS-Nummer: 1827-27-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR