(3,5-Dichlorphenyl)methansulfonsäurechlorid

Lieferant: SIA ENAMINE

Synonyme: 3,5-Dichloro-α-toluenesulphonyl chloride

EN300104226.1G
ENAMEN300104226.1GEA 0 EUR
ENAMEN300104226.1G
(3,5-Dichlorphenyl)methansulfonsäurechlorid
(3,5-Dichlorophenyl)methanesulphonyl chloride
Formel: C₇H₅CI₃O₂S
Molecular Weight: 259.54 g/mol
MDL: MFCD01631928
CAS-Nummer: 163295-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR