4-Chlor-m-toluylsäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 4-Chloro-3-methylbenzoic acid

EN300104262.1G
ENAMEN300104262.1GEA 0 EUR
ENAMEN300104262.1G
4-Chlor-m-toluylsäure
4-Chlor-m-toluylsäure
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
MDL: MFCD00045853
CAS-Nummer: 7697-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR