2-Fluor-3-hydroxypyridin

Lieferant: SIA ENAMINE

Synonyme: 2-Fluor-3-pyridinol

EN300104296.1G
ENAMEN300104296.1GEA 0 EUR
ENAMEN300104296.1G
2-Fluor-3-hydroxypyridin
2-Fluor-3-hydroxypyridin
Formel: C₅H₄FNO
Molecular Weight: 113,09 g/mol
MDL: MFCD04112569
CAS-Nummer: 174669-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR