3-Chlor-4-fluorbenzoesäure
Lieferant: SIA ENAMINE
EN300104562.1G
ENAMEN300104562.1GEA
0
EUR
ENAMEN300104562.1G
3-Chlor-4-fluorbenzoesäure
3-Chloro-4-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174,56 g/mol Siedepunkt: 273 °C (1013 hPa) Schmelzpunkt: 136 °C |
MDL:
MFCD00042477 CAS-Nummer: 403-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...