5-Brom-1,3-benzodioxol

Lieferant: SIA ENAMINE

Synonyme: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

EN300104950.1G
ENAMEN300104950.1GEA 0 EUR
ENAMEN300104950.1G
5-Brom-1,3-benzodioxol
5-Brom-1,3-benzodioxol
Formel: C₇H₅BrO₂
Molecular Weight: 201,02 g/mol
MDL: MFCD00005821
CAS-Nummer: 2635-13-4
EINECS: 220-123-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR