6-Bromchinaldin

Lieferant: SIA ENAMINE

EN300104966.1G
ENAMEN300104966.1GEA 0 EUR
ENAMEN300104966.1G
6-Bromchinaldin
6-Bromchinaldin
Formel: C₁₀H₈BrN
MDL: MFCD00079724
CAS-Nummer: 877-42-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR