3-Bromphenetol

Lieferant: SIA ENAMINE

Synonyme: 1-Brom-3-ethoxybenzol, (3-Bromphenyl)ethylether

EN300105078.1G
ENAMEN300105078.1GEA 0 EUR
ENAMEN300105078.1G
3-Bromphenetol
3-Bromphenetol
Formel: C₈H₉BrO
Molecular Weight: 201.06 g/mol
MDL: MFCD00274287
CAS-Nummer: 2655-84-7
EINECS: 220-186-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR