2-Chlor-6-nitrobenzonitril

Lieferant: SIA ENAMINE

Synonyme: 6-Chloro-2-nitrobenzonitrile

EN300105133.1G
ENAMEN300105133.1GEA 0 EUR
ENAMEN300105133.1G
2-Chlor-6-nitrobenzonitril
2-Chlor-6-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,57 g/mol
MDL: MFCD00007203
CAS-Nummer: 6575-07-1
EINECS: 229-496-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR