2-Methyl-5-nitrobenzotrifluorid

Lieferant: SIA ENAMINE

Synonyme: 1-Methyl-4-nitro-2-(trifluoromethyl)benzene

EN300105228.1G
ENAMEN300105228.1GEA 0 EUR
ENAMEN300105228.1G
2-Methyl-5-nitrobenzotrifluorid
2-Methyl-5-nitrobenzotrifluorid
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD01631684
CAS-Nummer: 89976-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR