D(-)-α-(4-Hydroxyphenyl)glycin

Lieferant: SIA ENAMINE

Synonyme: D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine

EN300105272.1G
ENAMEN300105272.1GEA 0 EUR
ENAMEN300105272.1G
D(-)-α-(4-Hydroxyphenyl)glycin
D(-)-α-(4-Hydroxyphenyl)glycin
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD00004262
CAS-Nummer: 22818-40-2
EINECS: 245-247-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR