1-Brom-4-chlor-2-fluorbenzol

Lieferant: SIA ENAMINE

EN300105273.1G
ENAMEN300105273.1GEA 0 EUR
ENAMEN300105273.1G
1-Brom-4-chlor-2-fluorbenzol
1-Brom-4-chlor-2-fluorbenzol
Formel: C₆H₃BrClF
Molecular Weight: 209,44 g/mol
MDL: MFCD00079708
CAS-Nummer: 1996-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR