(1S)-(-)-Borneol

Lieferant: SIA ENAMINE

Synonyme: L(-)-Borneol, (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanol

EN300105274.1G
ENAMEN300105274.1GEA 0 EUR
ENAMEN300105274.1G
(1S)-(-)-Borneol
(1S)-(-)-Borneol
Formel: C₁₀H₁₈O
Molecular Weight: 154,25 g/mol
Siedepunkt: 203 °C (1013 hPa)
Dichte: 1,011 g/cm³ (20 °C)
MDL: MFCD00003759
CAS-Nummer: 464-45-9
EINECS: 207-353-1
UN: 1312
ADR: 4.1,III
Merck Index: 13,01328

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR