2-(Cyclopentylmethoxy)benzonitril

Lieferant: SIA ENAMINE

EN300105282.1G
ENAMEN300105282.1GEA 0 EUR
ENAMEN300105282.1G
2-(Cyclopentylmethoxy)benzonitril
2-(Cyclopentylmethoxy)benzonitril
Formel: C₁₃H₁₅NO
MDL: MFCD14694456
CAS-Nummer: 1249696-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR