2-Brom-6-methoxyphenol

Lieferant: SIA ENAMINE

EN300105384.1G
ENAMEN300105384.1GEA 0 EUR
ENAMEN300105384.1G
2-Brom-6-methoxyphenol
2-Brom-6-methoxyphenol
Formel: C₇H₇BrO₂
MDL: MFCD08146628
CAS-Nummer: 28165-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR