α,α,α,5-Tetrafluor-m-toluylsäure

Lieferant: SIA ENAMINE

Synonyme: 3-Fluor-5-(trifluormethyl)benzoesäure

EN300105387.1G
ENAMEN300105387.1GEA 0 EUR
ENAMEN300105387.1G
α,α,α,5-Tetrafluor-m-toluylsäure
α,α,α,5-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
MDL: MFCD00061293
CAS-Nummer: 161622-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR