Ethyl-2-hydroxyisobutyrat

Lieferant: SIA ENAMINE

EN300105389.1G
ENAMEN300105389.1GEA 0 EUR
ENAMEN300105389.1G
Ethyl-2-hydroxyisobutyrat
Ethyl-2-hydroxyisobutyrat
Formel: C₆H₁₂O₃
Molecular Weight: 132,16 g/mol
MDL: MFCD00004458
CAS-Nummer: 80-55-7
EINECS: 201-290-3
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR