4-Brom-3-chlor-2-nitroanilin

Lieferant: SIA ENAMINE

EN300105464.1G
ENAMEN300105464.1GEA 0 EUR
ENAMEN300105464.1G
4-Brom-3-chlor-2-nitroanilin
4-Brom-3-chlor-2-nitroanilin
Formel: C₆H₄BrClN₂O₂
Molecular Weight: 251,4652 g/mol
MDL: MFCD09878187
CAS-Nummer: 1000573-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR