1-Brom-2-isopropoxybenzol

Lieferant: SIA ENAMINE

EN300105491.1G
ENAMEN300105491.1GEA 0 EUR
ENAMEN300105491.1G
1-Brom-2-isopropoxybenzol
1-Brom-2-isopropoxybenzol
Formel: C₉H₁₁BrO
MDL: MFCD00070759
CAS-Nummer: 701-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR