1-Brom-3-isopropoxybenzol

Lieferant: SIA ENAMINE

EN300105493.1G
ENAMEN300105493.1GEA 0 EUR
ENAMEN300105493.1G
1-Brom-3-isopropoxybenzol
1-Brom-3-isopropoxybenzol
Formel: C₉H₁₁BrO
MDL: MFCD00070756
CAS-Nummer: 131738-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR