2-Brom-5-chloranilin

Lieferant: SIA ENAMINE

EN300105774.1G
ENAMEN300105774.1GEA 0 EUR
ENAMEN300105774.1G
2-Brom-5-chloranilin
2-Brom-5-chloranilin
Formel: C₆H₅BrClN
MDL: MFCD00672942
CAS-Nummer: 823-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR