4-Brom-3-chloranilin

Lieferant: SIA ENAMINE

EN300105785.1G
ENAMEN300105785.1GEA 0 EUR
ENAMEN300105785.1G
4-Brom-3-chloranilin
4-Brom-3-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206,47 g/mol
Siedepunkt: 264 °C (1013 hPa)
Schmelzpunkt: 67…68 °C
MDL: MFCD00041311
CAS-Nummer: 21402-26-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR