4-Brom-3-chlorbenzoesäure

Lieferant: SIA ENAMINE
Achtung

EN300105786.1G
ENAMEN300105786.1GEA 0 EUR
ENAMEN300105786.1G
4-Brom-3-chlorbenzoesäure
4-Bromo-3-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD08276864
CAS-Nummer: 25118-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR