Methyl-5-hydroxynicotinat

Lieferant: SIA ENAMINE

Synonyme: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

EN300105837.1G
ENAMEN300105837.1GEA 0 EUR
ENAMEN300105837.1G
Methyl-5-hydroxynicotinat
Methyl-5-hydroxynicotinat
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
MDL: MFCD00191521
CAS-Nummer: 30766-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR