6-Chlor-5-nitronicotinsäure

Lieferant: SIA ENAMINE

EN300106043.1G
ENAMEN300106043.1GEA 0 EUR
ENAMEN300106043.1G
6-Chlor-5-nitronicotinsäure
6-Chlor-5-nitronicotinsäure
Formel: C₆H₃ClN₂O₄
MDL: MFCD09027297
CAS-Nummer: 7477-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR