3-Brom-4,5-dihydroxybenzaldehyd

Lieferant: SIA ENAMINE

EN300106071.1G
ENAMEN300106071.1GEA 0 EUR
ENAMEN300106071.1G
3-Brom-4,5-dihydroxybenzaldehyd
3-Bromo-4,5-dihydroxybenzaldehyde
Formel: C₇H₅BrO₃
MDL: MFCD00016608
CAS-Nummer: 16414-34-9
EINECS: 240-463-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR