4-(1-Pyrrolidinyl)anilin Hydrochlorid

Lieferant: SIA ENAMINE

EN30010626.1G
ENAMEN30010626.1GEA 0 EUR
ENAMEN30010626.1G
4-(1-Pyrrolidinyl)anilin Hydrochlorid
4-(1-Pyrrolidinyl)anilin Hydrochlorid
Formel: C₁₀H₁₅ClN₂
MDL: MFCD07700915
CAS-Nummer: 216670-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR