(4-Nitrophenoxy)essigsäure

Lieferant: SIA ENAMINE

Synonyme: 2-(4-Nitrophenoxy)acetic acid, 4-Nitrophenoxyacetic acid

EN30010631.1G
ENAMEN30010631.1GEA 0 EUR
ENAMEN30010631.1G
(4-Nitrophenoxy)essigsäure
(4-Nitrophenoxy)essigsäure
Formel: (C₆H₄(NO₂))OCH₂COOH
Molecular Weight: 197.15 g/mol
MDL: MFCD00017030
CAS-Nummer: 1798-11-4
EINECS: 217-283-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR