5'-Brom-3'-chlor-2'-hydroxyacetophenon
Lieferant: SIA ENAMINE
Synonyme:
1-(5-Bromo-3-chloro-2-hydroxyphenyl)ethan-1-one
EN300106332.1G
ENAMEN300106332.1GEA
0
EUR
ENAMEN300106332.1G
5'-Brom-3'-chlor-2'-hydroxyacetophenon
5'-Brom-3'-chlor-2'-hydroxyacetophenon
Formel:
C₈H₆BrClO₂ Molecular Weight: 249,489 g/mol |
MDL:
MFCD02660600 CAS-Nummer: 331821-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...