2-Hydrazino-3-nitropyridin

Lieferant: SIA ENAMINE

EN300106787.1G
ENAMEN300106787.1GEA 0 EUR
ENAMEN300106787.1G
2-Hydrazino-3-nitropyridin
2-Hydrazino-3-nitropyridin
Formel: C₅H₆N₄O₂
MDL: MFCD00085923
CAS-Nummer: 15367-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR