4-Brom-2-methylbenzaldehyd

Lieferant: SIA ENAMINE

EN300106816.1G
ENAMEN300106816.1GEA 0 EUR
ENAMEN300106816.1G
4-Brom-2-methylbenzaldehyd
4-Bromo-2-methylbenzaldehyde
Formel: C₈H₇BrO
MDL: MFCD07787171
CAS-Nummer: 24078-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR